missing translation for 'onlineSavingsMsg'
Learn More

Darovasertib, MedChemExpress

Product Code. 16440056
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
2mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16440056

Brand: Medchem Express HY101569

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Darovasertib (LXS196) is a potent, selective and orally active protein kinase C (PKC) inhibitor, with IC50 values of 1.9 nM, 0.4 nM and 3.1 μM for PKCα, PKCθ and GSK3β, respectively. Darovasertib has the potential for uveal melanoma research.

Darovasertib (LXS196) is a potent, selective and orally active protein kinase C (PKC) inhibitor, with IC50 values of 1.9 nM, 0.4 nM and 3.1 μM for PKCα, PKCθ and GSK3β, respectively. Darovasertib has the potential for uveal melanoma research.

Specifications

Chemical Name or Material Darovasertib
CAS 1874276-76-2
Color Yellow
Purity Grade Notes Research
Molecular Formula C22H23F3N8O
Quantity 2 mg
Synonym LXS196 IDE196
Solubility Information DMSO : 25 mg/mL (52.91 mM; Need ultrasonic)
SMILES NC1=C(C(NC2=C(N3CCC(C)(N)CC3)C=CC=N2)=O)N=C(C4=NC=CC=C4C(F)(F)F)C=N1
Molecular Weight (g/mol) 472.47
Formula Weight 472.47
Percent Purity 99.12%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.