missing translation for 'onlineSavingsMsg'
Learn More

Darovasertib, MedChemExpress

Product Code. 16474646
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
2mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16474646 100 mg 100mg
16466147 5 mg 5mg
16459946 50 mg 50mg
16440056 2 mg 2mg
16403428 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16474646 Supplier Medchem Express Supplier No. HY101569

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Darovasertib (LXS196) is a potent, selective and orally active protein kinase C (PKC) inhibitor, with IC50 values of 1.9 nM, 0.4 nM and 3.1 μM for PKCα, PKCθ and GSK3β, respectively. Darovasertib has the potential for uveal melanoma research.

Darovasertib (LXS196) is a potent, selective and orally active protein kinase C (PKC) inhibitor, with IC50 values of 1.9 nM, 0.4 nM and 3.1 μM for PKCα, PKCθ and GSK3β, respectively. Darovasertib has the potential for uveal melanoma research.

Specifications

Chemical Name or Material Darovasertib
CAS 1874276-76-2
Color Yellow
Purity Grade Notes Research
Molecular Formula C22H23F3N8O
Quantity 100 mg
Synonym LXS196 IDE196
Solubility Information DMSO : 25 mg/mL (52.91 mM; Need ultrasonic)
SMILES NC1=C(C(NC2=C(N3CCC(C)(N)CC3)C=CC=N2)=O)N=C(C4=NC=CC=C4C(F)(F)F)C=N1
Molecular Weight (g/mol) 472.47
Formula Weight 472.47
Percent Purity 99.12%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.