missing translation for 'onlineSavingsMsg'
Learn More

Darolutamide, MedChemExpress

Product Code. 16487206
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. Quantity unitSize
16487206 100 mg 100mg
16467238 10 mg 10mg
16449767 5 mg 5mg
16433257 50 mg 50mg
4 options
This item is not returnable. View return policy

Product Code. 16487206

Brand: Medchem Express HY16985

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Darolutamide (ODM-201;BAY-1841788) is a potent androgen receptor (AR) antagonist with an IC50 of 26 nM in in vitro assay.

Darolutamide (ODM-201

Specifications

Chemical Name or Material Darolutamide
Physical Form Powder
CAS 1297538-32-9
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C19H19ClN6O2
For Use With (Application) Cancer-programmed cell death
Synonym ODM-201 BAY-1841788
Solubility Information DMSO : 125 mg/mL (313.40 mM; Need ultrasonic)
SMILES ClC1=C(C#N)C=CC(C2=NN(C[C@H](C)NC(C3=NNC(C(O)C)=C3)=O)C=C2)=C1
Molecular Weight (g/mol) 398.85
Formula Weight 398.85
Percent Purity 98.1%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.