missing translation for 'onlineSavingsMsg'
Learn More

Darodipine, TRC

Product Code. 30136100
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136100 25 mg 25mg
30005905 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30136100 Supplier Toronto Research Chemicals Supplier No. TRCD19345025MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72803-02-2
Molecular Formula C19 H21 N3 O5
Molecular Weight (g/mol) 371.39
Synonym 4-(2,1,3-Benzoxadiazol-4-yl)-1,4-dihydro-2,6-dimethyl-3,5-pyridinedicarboxylic Acid 3,5-Diethyl Ester, 2,1,3-Benzoxadiazole, 3,5-pyridinedicarboxylic Acid deriv., PY 108-068, PY 108068;
IUPAC Name diethyl 4-(2,1,3-benzoxadiazol-4-yl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate
SMILES CCOC(=O)C1=C(C)NC(=C(C1c2cccc3nonc23)C(=O)OCC)C

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C19H21N3O5/c1-5-25-18(23)14-10(3)20-11(4)15(19(24)26-6-2)16(14)12-8-7-9-13-17(12)22-27-21-13/h7-9,16,20H,5-6H2,1-4H3
Formula Weight 371.1481
Chemical Name or Material Darodipine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.