missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DARC Chimeric Recombinant Rabbit Monoclonal Antibody (2C3)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA548105
This item is not returnable.
View return policy
Description
Specificity: The antibody binds specifcally to the pentapeptide FEDVW of the Duffy receptor.
ACKR1 or DARC is an atypical chemokine receptor that controls chemokine levels and localization via high-affinity chemokine binding that is uncoupled from classic ligand-driven signal transduction cascades resulting instead in chemokine sequestration, degradation, or transcytosis. ACKR1 is also known as an interceptor (internalizing receptor) or chemokine-scavenging receptor or chemokine decoy receptor. It has a promiscuous chemokine-binding profile by interacting with inflammatory chemokines of both the CXC and the CC subfamilies but not with homeostatic chemokines. ACKR1 also acts as a receptor for chemokines including CCL2, CCL5, CCL7, CCL11, CCL13, CCL14, CCL17, CXCL5, CXCL6, IL8/CXCL8, CXCL11, GRO, RANTES, MCP-1, TARC and for the malaria parasites P.vivax and P.knowlesi.
Specifications
| DARC Chimeric | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.02% ProClin 300 | |
| Q16570 | |
| ACKR1 | |
| CHO cells transfected with cDNA encoding the Duffy glycoprotein. | |
| 200 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG κ |
| ELISA, Flow Cytometry, Inhibition Assays, Western Blot, Immunocytochemistry | |
| 2C3 | |
| Unconjugated | |
| ACKR1 | |
| AA162249; ACKR1; Atypical chemokine receptor 1; atypical chemokine receptor 1 (Duffy blood group); CCBP1; CD234; CD234 antigen; CTA-134P22.3; DARC; Dfy; Duffy Antigen; Duffy antigen chemokine receptor; Duffy antigen receptor for chemokines; duffy antigen/chemokine receptor; Duffy blood group antigen; duffy blood group chemokine receptor; Duffy blood group, atypical chemokine receptor; Duffy blood group, chemokine receptor; ESTM35; FY; Fy Antigen; Fy glycoprotein; FYDuffy blood group; glycoprotein D; GPD; GPDWBCQ1; GpFy; HGNC:4035; OTTHUMP00000060081; Plasmodium vivax receptor; WBCQ1 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 2532 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction