missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
DARC Monoclonal antibody specifically detects DARC in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF
Specifications
Specifications
| Antigen | DARC |
| Applications | Flow Cytometry, Immunocytochemistry, CyTOF |
| Classification | Monoclonal |
| Clone | 358307 |
| Conjugate | mFluor Violet 450 SE |
| Dilution | Flow Cytometry, Immunocytochemistry, CyTOF-ready |
| Formulation | 50mM Sodium Borate |
| Gene Alias | CCBP1, CD234, CD234 antigen, Dfy, Duffy antigen/chemokine receptor, Duffy blood group antigen, Duffy blood group, chemokine receptor, Fy glycoprotein, FYDuffy blood group, Glycoprotein D, GPDWBCQ1, GpFy, Plasmodium vivax receptor |
| Host Species | Mouse |
| Immunogen | HEK293 human embryonic kidney cell line transfected with human DARC, Met1-Ser336, Accession # Q16570 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?