missing translation for 'onlineSavingsMsg'
Learn More

DARC Antibody (2C3) - Chimeric - Azide and BSA Free, Novus Biologicals™

Product Code. 18387041 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.2 mg
Unit Size:
0.20mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18387041 0.2 mg 0.20mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18387041 Supplier Novus Biologicals Supplier No. NBP275197

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

DARC Monoclonal specifically detects DARC in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunofluorescence, Block/Neutralize.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DARC
Applications Western Blot, Flow Cytometry, ELISA, Immunofluorescence, Blocking Assay
Classification Monoclonal
Clone 2C3
Conjugate Unconjugated
Dilution Western Blot, Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunofluorescence, Block/Neutralize
Formulation PBS with 0.02% Proclin 300
Gene Alias CCBP1, CD234, CD234 antigen, Dfy, Duffy antigen/chemokine receptor, Duffy blood group antigen, Duffy blood group, chemokine receptor, Fy glycoprotein, FYDuffy blood group, Glycoprotein D, GPDWBCQ1, GpFy, Plasmodium vivax receptor
Gene Symbols ACKR1
Host Species Rabbit
Immunogen CHO cells transfected with cDNA encoding the Duffy glycoprotein.
Purification Method Protein A purified
Quantity 0.2 mg
Primary or Secondary Primary
Gene ID (Entrez) 2532.0
Test Specificity The antibody binds specifcally to the pentapeptide FEDVW of the Duffy receptor.
Target Species Human
Content And Storage Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.