missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Daptomycin Impurity B1-I, TRC
Specifications
Specifications
| Chemical Name or Material | Daptomycin Impurity B1-I |
| Analyte or Component Names | Daptomycin Impurity B1-I |
| Shipping Condition | Room Temperature |
| Molecular Formula | C44H66N10O15 • (C2HF3O2) |
| Synonym | Decanoyl-Trp-(D-Asn)-Asp-Thr-Gly-Orn-Asp-OH Trifluoroacetic Acid Salt, (5S,14S)-14-((R)-4-Amino-2-((S)-2-decanamido-3-(1H-indol-3-yl)propanamido)-4-oxobutanamido)-5-(3-aminopropyl)-11-((R)-1-hydroxyethyl)-4,7,10,13-tetraoxo-3,6,9,12-tetraazapentadecane-1,2,15-tricarboxylic Acid Trifluoroacetic Acid Salt |
| InChI Formula | InChI=1S/C44H66N10O15.CHF3.CH2O2/c1-3-4-5-6-7-8-9-16-34(57)50-29(18-25-22-47-27-14-11-10-13-26(25)27)40(64)51-30(19-33(46)56)41(65)52-31(20-36(59)60)42(66)54-38(24(2)55)43(67)48-23-35(58)49-28(15-12-17-45)39(63)53-32(44(68)69)21-37(61)62;2-1(3)4;2-1-3/h10-11,13-14,22,24,28-32,38,47,55H,3-9,12,15-21,23,45H2,1-2H3,(H2,46,56)(H,48,67)(H,49,58)(H,50,57)(H,51,64)(H,52,65)(H,53,63)(H,54,66)(H,59,60)(H,61,62)(H,68,69);1H;1H,(H,2,3)/t24?,28-,29-,30+,31-,32?,38?;;/m0../s1 |
| Solution Type | Neat |
| SMILES | O=C(CCCCCCCCC)N[C@H](C(N[C@H](CC(N)=O)C(N[C@@H](CC(O)=O)C(NC(C(NCC(N[C@@H](CCCN)C(NC(CC(O)=O)C(O)=O)=O)=O)=O)C(O)C)=O)=O)=O)CC1=CNC2=C1C=CC=C2.F[C](F)F.O=CO |
| IUPAC Name | 2-[[(2S)-5-amino-2-[[2-[[2-[[(2S)-2-[[(2R)-4-amino-2-[[(2S)-2-(decanoylamino)-3-(1H-indol-3-yl)propanoyl]amino]-4-oxobutanoyl]amino]-3-carboxypropanoyl]amino]-3-hydroxybutanoyl]amino]acetyl]amino]pentanoyl]amino]butanedioic acid;fluoroform;formic acid |
| Industry Type | Pharmaceutical |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?