missing translation for 'onlineSavingsMsg'
Learn More

DAPT, MedChemExpress

Product Code. p-200098977
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Product Code. Quantity unitSize
16478207 5 mg 5mg
16435498 10 mg 10mg
16491667 50 mg 50mg
3 options
This item is not returnable. View return policy

Product Code. 16478207

Brand: Medchem Express HY13027

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress DAPT (GSI-IX) is a potent and orally active γ-secretase inhibitor with IC50s of 115 nM and 200 nM for total amyloid-β (Aβ) and Aβ42, respectively.

DAPT inhibits the activation of Notch 1 signaling and induces cell differentiation. DAPT also induces autophagy and apoptosis. DAPT has neuroprotection activity and has the potential for autoimmune and lymphoproliferative diseases, degenerative disease and cancers treatment.

Specifications

Chemical Name or Material DAPT
CAS 208255-80-5
Color White
Purity Grade Notes Research
Molecular Formula C23H26F2N2O4
Quantity 5 mg
Synonym GSI-IX
Solubility Information DMSO : 62.5 mg/mL (144.52 mM; Need ultrasonic) ∣Ethanol : 10 mg/mL (23.12 mM; Need ultrasonic)
SMILES FC1=CC(F)=CC(CC(N[C@H](C(N[C@H](C(OC(C)(C)C)=O)C2=CC=CC=C2)=O)C)=O)=C1
Molecular Weight (g/mol) 432.46
Formula Weight 432.46
Percent Purity 99.62%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.