missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DAPT, MedChemExpress
Description
DAPT inhibits the activation of Notch 1 signaling and induces cell differentiation. DAPT also induces autophagy and apoptosis. DAPT has neuroprotection activity and has the potential for autoimmune and lymphoproliferative diseases, degenerative disease and cancers treatment.
Specifications
Specifications
| Chemical Name or Material | DAPT |
| CAS | 208255-80-5 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C23H26F2N2O4 |
| Quantity | 5 mg |
| Synonym | GSI-IX |
| Solubility Information | DMSO : 62.5 mg/mL (144.52 mM; Need ultrasonic) ∣Ethanol : 10 mg/mL (23.12 mM; Need ultrasonic) |
| SMILES | FC1=CC(F)=CC(CC(N[C@H](C(N[C@H](C(OC(C)(C)C)=O)C2=CC=CC=C2)=O)C)=O)=C1 |
| Molecular Weight (g/mol) | 432.46 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?