missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies DAPT
Description
- DAPT is an inhibitor of the γ-secretase complex
- Notch is a key target of γ-secretase, therefore DAPT indirectly inhibits the Notch pathway
- Other targets of γ-secretase that would be influenced by DAPT include amyloid precursor protein, E-cadherin, and ErbB4
Specifications
Specifications
| CAS | 208255-80-5 |
| Synonym | GSI-IX; LY-374973 |
| Quantity | 5 mg |
| Molecular Formula | C23H26F2N2O4 |
| Purity | ≥95% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?