missing translation for 'onlineSavingsMsg'
Learn More

Dapsone, MedChemExpress

Product Code. 16462259
Encompass_Preferred
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 16462259

Brand: Medchem Express HYB0688

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Dapsone (4,4′-Diaminodiphenyl sulfone) is an orally active and blood-brain penetrant sulfonamide antibiotic with bacteriostatic, antimycobacterial and antiprotozoal activities. Dapsone exerts effective antileprosy activity and inhibits folate synthesis in cell extracts of M. leprae. Dapsone is used for dermatologic disorder research, including leprosy, dermatitis herpetiformis, acne vulgaris et al.

Dapsone (4,4′-Diaminodiphenyl sulfone) is an orally active and blood-brain penetrant sulfonamide antibiotic with bacteriostatic, antimycobacterial and antiprotozoal activities. Dapsone exerts effective antileprosy activity and inhibits folate synthesis in cell extracts of M. leprae. Dapsone is used for dermatologic disorder research, including leprosy, dermatitis herpetiformis, acne vulgaris et al.

Specifications

Chemical Name or Material Dapsone
Physical Form Powder
CAS 80-08-0
Color White
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C12H12N2O2S
For Use With (Application) COVID-19-immunoregulation
Synonym 4,4′-Diaminodiphenyl sulfone DDS
Solubility Information DMSO : 100 mg/mL (402.74 mM; Need ultrasonic)
SMILES O=S(C1=CC=C(N)C=C1)(C2=CC=C(N)C=C2)=O
Molecular Weight (g/mol) 248.3
Formula Weight 248.3
Percent Purity 98.7%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.