missing translation for 'onlineSavingsMsg'
Learn More

Dapsone, MedChemExpress

Product Code. 16491038
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16491038 5 g 5g
16462259 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16491038 Supplier Medchem Express Supplier No. HYB0688
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Dapsone (4,4′-Diaminodiphenyl sulfone) is an orally active and blood-brain penetrant sulfonamide antibiotic with bacteriostatic, antimycobacterial and antiprotozoal activities. Dapsone exerts effective antileprosy activity and inhibits folate synthesis in cell extracts of M. leprae. Dapsone is used for dermatologic disorder research, including leprosy, dermatitis herpetiformis, acne vulgaris et al.

Dapsone (4,4′-Diaminodiphenyl sulfone) is an orally active and blood-brain penetrant sulfonamide antibiotic with bacteriostatic, antimycobacterial and antiprotozoal activities. Dapsone exerts effective antileprosy activity and inhibits folate synthesis in cell extracts of M. leprae. Dapsone is used for dermatologic disorder research, including leprosy, dermatitis herpetiformis, acne vulgaris et al.

Specifications

Chemical Name or Material Dapsone
Physical Form Powder
CAS 80-08-0
Color White
Quantity 5 g
Purity Grade Notes Research
Molecular Formula C12H12N2O2S
For Use With (Application) COVID-19-immunoregulation
Synonym 4,4′-Diaminodiphenyl sulfone DDS
Solubility Information DMSO : 100 mg/mL (402.74 mM; Need ultrasonic)
SMILES O=S(C1=CC=C(N)C=C1)(C2=CC=C(N)C=C2)=O
Molecular Weight (g/mol) 248.3
Formula Weight 248.3
Percent Purity 98.7%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.