missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DAPP1 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5141982
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:4,000.
May act as a B-cell-associated adapter that regulates B-cell antigen receptor (BCR)-signaling downstream of PI3K.
Specifications
| DAPP1 | |
| Polyclonal | |
| Unconjugated | |
| DAPP1 | |
| b lymphocyte adapter protein Bam32; Bam32; B-cell adapter molecule of 32 kDa; Dapp1; Dual adapter for phosphotyrosine and 3-phosphotyrosine and 3-phosphoinositide; dual adaptor for phosphotyrosine and 3-phosphoinositides 1; dual adaptor of phosphotyrosine and 3-phosphoinositides; dual adaptor of phosphotyrosine and 3-phosphoinositides 1; hDAPP1; HSPC066; mDAPP1 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 27071 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q9UN19 | |
| DAPP1 | |
| Peptide with sequence SALCISPEEKTDHK (aa 267-280). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction