missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DAPP1 Mouse anti-Human, Clone:2F7A9, Abnova™
Mouse monoclonal antibody raised against recombinant human DAPP1.
Brand: Abnova MAB17230.100ug
This item is not returnable.
View return policy
Specifications
| DAPP1 | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Western Blot (1:500-1:2000) Immunohistochemistry Immunocytochemistry Flow Cytometry (1:200-1:400) The optimal working dilution should be determined by the end user. |
|
| DAPP1 | |
| Mouse | |
| 100 μg | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Western Blot | |
| 2F7A9 | |
| dual adaptor of phosphotyrosine and 3-phosphoinositides | |
| In PBS (0.05% sodium azide) | |
| BAM32, DKFZp667E0716 | |
| Recombinant protein corresponding to amino acid 127-276 of human DAPP1 from E. coli. | |
| 27071 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction