missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
DAPP1 Polyclonal specifically detects DAPP1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
| Antigen | DAPP1 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Formulation | PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide |
| Gene Accession No. | Q9UN19 |
| Gene Alias | B lymphocyte adapter protein Bam32, BAM32DKFZp667E0716, B-cell adapter molecule of 32 kDa, dual adapter for phosphotyrosine and 3-phosphotyrosine and 3-phosphoinositide, dual adaptor of phosphotyrosine and 3-phosphoinositides, hDAPP1 |
| Gene Symbols | DAPP1 |
| Host Species | Rabbit |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?