missing translation for 'onlineSavingsMsg'
Learn More

DAPP1 Antibody [DyLight 755], Novus Biologicals™

Product Code. 30734096 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30734096 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30734096 Supplier Novus Biologicals Supplier No. AF7024Z

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

DAPP1 Polyclonal antibody specifically detects DAPP1 in Human,Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen DAPP1
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias B lymphocyte adapter protein Bam32, BAM32DKFZp667E0716, B-cell adapter molecule of 32 kDa, dual adapter for phosphotyrosine and 3-phosphotyrosine and 3-phosphoinositide, dual adaptor of phosphotyrosine and 3-phosphoinositides, hDAPP1
Host Species Sheep
Immunogen E. coli-derived recombinant human DAPP1, Gly2-Ser163, Accession # Q9UN19
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 27071
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.