missing translation for 'onlineSavingsMsg'
Learn More

Dapoxetine Hydrochloride, TRC

Product Code. 17974005
Change view
Click to view available options
Quantity:
10 mg
100 mg
500 mg
Unit Size:
100mg
10mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17974005 500 mg 500mg
17904885 10 mg 10mg
17914145 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17974005 Supplier Toronto Research Chemicals Supplier No. TRCD185700500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 129938-20-1
Molecular Formula C21 H23 N O . Cl H
Molecular Weight (g/mol) 341.87
Synonym (αS)-N,N-Dimethyl-α-[2-(1-naphthalenyloxy)ethyl]-benzenemethanamine Hydrochloride (1:1), Benzenemethanamine, N,N-dimethyl-α-[2-(1-naphthalenyloxy)ethyl]-, hydrochloride, (S)-, Benzenemethanamine, N,N-dimethyl-α-[2-(1-naphthalenyloxy)ethyl]-, hydrochloride, (αS)- (9CI), (S)-(+)-Dapoxetine hydrochloride, Dapoxetine hydrochloride, LY 210448 hydrochloride, Priligy
IUPAC Name (1S)-N,N-dimethyl-3-naphthalen-1-yloxy-1-phenylpropan-1-amine;hydrochloride
SMILES Cl.CN(C)[C@@H](CCOc1cccc2ccccc12)c3ccccc3

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C21H23NO.ClH/c1-22(2)20(18-10-4-3-5-11-18)15-16-23-21-14-8-12-17-9-6-7-13-19(17)21;/h3-14,20H,15-16H2,1-2H3;1H/t20-;/m0./s1
Formula Weight 341.1546
Percent Purity >95%
Chemical Name or Material Dapoxetine Hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.