Learn More
Invitrogen™ Human DAPK3 (ZIPK), GST Tag Recombinant Protein
Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length
Specifications
Specifications
| Accession Number | O43293 |
| Concentration | See Label |
| For Use With (Application) | Kinase Assay |
| Formulation | 50 mM tris with 0.02% Triton X-100, 0.5 mM EDTA, 150 mM NaCl, 2 mM DTT, 50% glycerol and no preservative; pH 7.5 |
| Gene ID (Entrez) | 1613 |
| Molecular Weight (g/mol) | 80 kDa |
| Name | Human DAPK3 (ZIPK), GST Tag |
| Quantity | 1 mg |
| Regulatory Status | RUO - research use only |
| Gene Alias | DAP kinase 3; DAP3K; Dapk3; Dapkl; DAP-like kinase; death associated protein kinase 3; death-associated kinase 3; Death-associated like kinase; death-associated protein kinase 3; DLK; MYPT1 kinase; ZIP; ZIP kinase; ZIP kinase isoform; Zipk; ZIP-kinase; zipper-interacting protein kinase |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.