Learn More
DAPK3 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 30656-1-AP-20UL
This item is not returnable.
View return policy
Description
The DAPK3 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human DAPK3. This antibody recognizes human antigen. The DAPK3 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| DAPK3 | |
| 500 μg/mL | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |
| Western Blot, ELISA | |
| DAP kinase 3, DAP like kinase, DAPK3, Dlk, FLJ36473, ZIP, ZIP kinase, ZIPK | |
| 20 μL | |
| 1613 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |