Learn More
Eurofins DiscoverX DAPK2 Protein, Active
Description
Death-associated protein kinase (DAPK) is a calcium/calmodulin-dependent kinase that phosphorylates serine/threonine residues on proteins. DAPK modulates cell death induced by IFN-g, TNF-a, Fas, and detachment from the extracellular matrix. DAPK has been also implicated in TGF-binduced apoptosis in several cultured cell lines in which Smad proteins mediate transcriptional activation of DAPK. A member of DAP kinase family, DAPK2 is a 42 KDa calcium/calmodulin-dependant serine/threonine kinase localized in the cytoplasm. Reports suggest DAPK2 along with DAPK1 induces membrane blebbing and formation of autophagic vesicles and plays a vital role in TGF-induced caspase-dependent Type-I and IFNg-induced caspase-independent autophagic type II cell death signaling. Inhibition of DAPK2 may facilitate tumor progression.
GenBank NM_014326
Specifications
Specifications
| Formulation | mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 300 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. |
| Molecular Weight (g/mol) | 70.6 kDa |
| Quantity | 10 μg |
| Species | Human |
| Recombinant | Recombinant |
| Protein Tag | N-terminal GST |
| Expression System | Recombinant enzyme expressed in Sf21 insect cells |
| Protein Form | Full-length |
| Purity or Quality Grade | ≥68% |
| Protein | DAPK2, active |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.