missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DAPK2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA141305
This item is not returnable.
View return policy
Description
Suggested positive control: antigen standard for DAPK2 (transient overexpression lysate), rat kidney tissue, rat kidney protein.
DAPK2 is a calcium/calmodulin-dependent kinase that acts as a positive regulator of apoptosis, and is expressed in many tissue types. DAPK2 encodes a protein that belongs to the serine/threonine protein kinase family. This protein contains a N-terminal protein kinase domain followed by a conserved calmodulin-binding domain with significant similarity to that of death-associated protein kinase 1 (DAPK1), a positive regulator of programmed cell death. Overexpression of this gene was shown to induce cell apoptosis. It uses multiple polyadenylation sites.
Specifications
| DAPK2 | |
| Polyclonal | |
| Unconjugated | |
| DAPK2 | |
| DAP kinase 2; DAPK2; DAP-kinase-related protein 1; DAP-kinase-related protein 1 beta isoform; death associated protein kinase 2; death-associated kinase 2; death-associated protein kinase 2; DRAK2; DRP1; DRP-1; MGC119312 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 13143, 23604, 300799 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.05% BSA and 0.05% sodium azide | |
| Q8VDF3, Q9UIK4 | |
| DAPK2 | |
| KLH-conjugated synthetic peptides corresponding to amino acids 325-342 and 347-364 of human DAPK2. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction