missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DAPK2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 12858172
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12858172 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12858172 Supplier Invitrogen™ Supplier No. PA519961

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This antibody will not cross-react with DAPK1. In Western blot applications, this antibody detects a band at ~42kDa. A suggested positive control is A431 cell lysate. PA5-19961 can be used with blocking peptide PEP-0086.

DAPK2 is a calcium/calmodulin-dependent kinase that acts as a positive regulator of apoptosis, and is expressed in many tissue types. DAPK2 encodes a protein that belongs to the serine/threonine protein kinase family. This protein contains a N-terminal protein kinase domain followed by a conserved calmodulin-binding domain with significant similarity to that of death-associated protein kinase 1 (DAPK1), a positive regulator of programmed cell death. Overexpression of this gene was shown to induce cell apoptosis. It uses multiple polyadenylation sites.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DAPK2
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene DAPK2
Gene Accession No. Q8VDF3, Q9UIK4
Gene Alias DAP kinase 2; DAPK2; DAP-kinase-related protein 1; DAP-kinase-related protein 1 beta isoform; death associated protein kinase 2; death-associated kinase 2; death-associated protein kinase 2; DRAK2; DRP1; DRP-1; MGC119312
Gene Symbols DAPK2
Host Species Rabbit
Immunogen A peptide corresponding to amino acids near the carboxy terminus of human DAPK2.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13143, 23604, 300799
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.