missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DAPK2 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA19035
This item is not returnable.
View return policy
Description
This antibody is predicted to react with canine, porcine and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.
DAPK2 is a calcium/calmodulin-dependent kinase that acts as a positive regulator of apoptosis, and is expressed in many tissue types. DAPK2 encodes a protein that belongs to the serine/threonine protein kinase family. This protein contains a N-terminal protein kinase domain followed by a conserved calmodulin-binding domain with significant similarity to that of death-associated protein kinase 1 (DAPK1), a positive regulator of programmed cell death. Overexpression of this gene was shown to induce cell apoptosis. It uses multiple polyadenylation sites.
Specifications
| DAPK2 | |
| Polyclonal | |
| Unconjugated | |
| DAPK2 | |
| DAP kinase 2; DAPK2; DAP-kinase-related protein 1; DAP-kinase-related protein 1 beta isoform; death associated protein kinase 2; death-associated kinase 2; death-associated protein kinase 2; DRAK2; DRP1; DRP-1; MGC119312 | |
| Goat | |
| Ammonium Sulfate Precipitation | |
| RUO | |
| 13143, 23604 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q8VDF3, Q9UIK4 | |
| DAPK2 | |
| Synthetic peptide corresponding to the C terminus amino acids KALHPRRRSSTS (aa 359-370). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction