missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ DAPI

Product Code. 16214970
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
This item is not returnable. View return policy

Product Code. 16214970

Brand: Abnova™ U0331.10mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

DAPI is a nuclear and chromosome counterstain, it emits blue fluorescence upon binding to AT regions of DNA. Although the dye is cell impermeant, higher concentrations will enter a live cell.

Specifications

Content And Storage Store at room temperature and protect from light.
For Use With (Application) Cell Staining
Quantity 10 mg
compliance-icons
Product Identifier
  • 10MG DAPI
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P233-Keep container tightly closed.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P404-Store in a closed container.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : sodium phosphate dibasic, potassium phosphate monobasic
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.