missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Daphnetin, MedChemExpress
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Description
Daphnetin triggers ROS-induced cell apoptosis and induces cytoprotective autophagy by modulating the AMPK/Akt/mTOR pathway. Daphnetin has anti-inflammation activitity and inhibits TNF-α, IL-1ß, ROS, and MDA production. Daphnetin has schizontocidal activity against malaria parasites. Daphnetin can be used for rheumatoid arthritis , cancer and anti-malarian research.
Specifications
Specifications
| Chemical Name or Material | Daphnetin |
| Physical Form | Solid |
| CAS | 486-35-1 |
| Color | Light Yellow |
| Quantity | 10 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C9H6O4 |
| For Use With (Application) | COVID-19-immunoregulation |
| Synonym | 7,8-Dihydroxycoumarin |
| Solubility Information | DMSO : 50 mg/mL (280.68 mM; Need ultrasonic) ∣H2O : 1 mg/mL (5.61 mM; Need ultrasonic) |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction