missing translation for 'onlineSavingsMsg'
Learn More

Dapagliflozin, MedChemExpress

Product Code. 16496687
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16496687

Brand: Medchem Express HY10450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Dapagliflozin (BMS-512148), a new type of drug used to treat diabetes mellitus (DM), is a competitive sodium/glucose cotransporter 2 (SGLT2) inhibitor, which results in excretion of glucose into the urine. Dapagliflozin induces HIF1 expression and attenuates renal IR injury.

Dapagliflozin (BMS-512148), a new type of drug used to treat diabetes mellitus (DM), is a competitive sodium/glucose cotransporter 2 (SGLT2) inhibitor, which results in excretion of glucose into the urine.

Specifications

Chemical Name or Material Dapagliflozin
CAS 461432-26-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C21H25ClO6
Quantity 5 mg
Synonym BMS-512148
Solubility Information DMSO : ≥ 100 mg/mL (244.58 mM) ∣H2O : 50 mg/mL (122.29 mM; Need ultrasonic)
SMILES O[C@H]1[C@H](C2=CC(CC3=CC=C(OCC)C=C3)=C(Cl)C=C2)O[C@H](CO)[C@@H](O)[C@@H]1O
Molecular Weight (g/mol) 408.87
Formula Weight 408.87
Percent Purity 95.0%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.