missing translation for 'onlineSavingsMsg'
Learn More

Dapagliflozin ((2S)-1,2-propanediol, hydrate), MedChemExpress

Product Code. 16473978
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16473978 10 mg 10mg
16454986 100 mg 100mg
16420337 50 mg 50mg
16406697 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16473978 Supplier Medchem Express Supplier No. HY10450A
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Dapagliflozin ((2S)-1,2-propanediol, hydrate) is the S-enantiomer of Dapagliflozin 1,2-propanediol, hydrate.

Dapagliflozin ((2S)-1,2-propanediol, hydrate), a new type of drug used to treat diabetes mellitus (DM), is a competitive sodium/glucose cotransporter 2 (SGLT2) inhibitor, which results in excretion of glucose into the urine. Dapagliflozin ((2S)-1,2-propanediol, hydrate) induces HIF1 expression and attenuates renal IR injury.

Specifications

Chemical Name or Material Dapagliflozin ((2S)-1,2-propanediol, hydrate)
CAS 960404-48-2
Color White
Purity Grade Notes Research
Molecular Formula C24H35ClO9
Quantity 10 mg
Synonym BMS-512148 (2S)-1,2-propanediol, hydrate
Solubility Information DMSO : ≥ 100 mg/mL (198.82 mM) ∣H2O : 2.38 mg/mL (4.73 mM; Need ultrasonic)
SMILES O[C@H]1[C@H](C2=CC(CC3=CC=C(OCC)C=C3)=C(Cl)C=C2)O[C@H](CO)[C@@H](O)[C@@H]1O.C[C@H](O)CO.O
Molecular Weight (g/mol) 502.98
Formula Weight 502.98
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.