missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DAP5 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15700301 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15700301 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15700301 Supplier Invitrogen™ Supplier No. PA552345

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: QDTVELREHH WVPRKAFLDN GPKTINQIRQ DAVKDLGVFI PAPMAQGMRS DFFLEGPFMP PRMKMDRDPL GGLADMFGQM PGSGIGTGPG VIQDRFSPTM GRHRSNQLFN GHGGHIMPPT QSQFGEMGGK FMKSQGLSQL YH Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.

CDK5RAP2 is a regulator of CDK5 (cyclin-dependent kinase 5) activity. The protein is localized to the centrosome and Golgi complex and interacts with CDK5R1 and pericentrin (PCNT). CDK5RAP2 plays a role in centriole engagement and microtubule nucleation, and has been linked to primary microcephaly and Alzheimer's disease. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DAP5
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Eif4g2
Gene Accession No. P78344, Q62448
Gene Alias AA589388; AAG1; aging-associated protein 1; DAP5; DAP-5; Death-associated protein 5; E130105L11Rik; eIF4G 2; eIF-4G 2; EIF4G2; eif4g2.L; eif4g2.S; eIF-4-gamma 2; Eukaryotic translation initiation factor 4 gamma 2; eukaryotic translation initiation factor 4 gamma, 2; eukaryotic translation initiation factor 4 gamma, 2 L homeolog; eukaryotic translation initiation factor 4 gamma, 2 S homeolog; eukaryotic translation initiation factor 4 gamma, 2 S homeolog; eukaryotic translation initiation factor 4, gamma 2; eukaryotic translation initiation factor 4G-like 1; FLJ41344; NAT1; Natm1; Novel APOBEC-1 target 1; OK/SW-cl.75; p97; translation repressor NAT1; XELAEV_18024574mg
Gene Symbols Eif4g2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DAP5. Recombinant protein control fragment (Product #RP-88745).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13690, 1982, 361628
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.