missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DAP5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38020-20ul
This item is not returnable.
View return policy
Description
DAP5 Polyclonal antibody specifically detects DAP5 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Immunoprecipitation,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifications
| DAP5 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunoprecipitation 0.5ug - 4ug antibody for 200ug - 400ug extracts of whole cells, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| aging-associated protein 1, DAP-5, DAP5AAG1, Death-associated protein 5, eIF4G 2, eIF-4G 2, eIF-4-gamma 2, eukaryotic translation initiation factor 4 gamma 2, eukaryotic translation initiation factor 4 gamma, 2, eukaryotic translation initiation factor 4G-like 1, FLJ41344, NAT1, p97 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 320-490 of human DAP5 (NP_001409.3).,, Sequence:, PKTINQIRQDAVKDLGVFIPAPMAQGMRSDFFLEGPFMPPRMKMDRDPLGGLADMFGQMPGSGIGTGPGVIQDRFSPTMGRHRSNQLFNGHGGHIMPPTQSQFGEMGGKFMKSQGLSQLYHNQSQGLLSQLQGQSKDMPPRFSKKGQLNADEISLRPAQSFLMNKNQVPKL | |
| 20 μL | |
| Apoptosis, mTOR Pathway | |
| 1982 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction