missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DAP3 Antibody (42C617.1.2), Novus Biologicals™
Mouse Monoclonal Antibody
£167.00 - £378.00
Specifications
| Antigen | DAP3 |
|---|---|
| Clone | 42C617.1.2 |
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 3-5 ug/ml |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18001454
|
Novus Biologicals
NB100-56539SS |
0.025 mg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18047985
|
Novus Biologicals
NB100-56539 |
0.1 mg |
£378.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DAP3 Monoclonal specifically detects DAP3 in Human, Mouse samples. It is validated for Western Blot.Specifications
| DAP3 | |
| 0.5 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Core ESC Like Genes, Stem Cell Markers | |
| P51398 | |
| 7818 | |
| The antibody was raised against a synthetic peptide corresponding to amino acids 51-66 of human DAP-3 protein. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 42C617.1.2 | |
| Western Blot 3-5 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| bMRP-10, DAP-3Death-associated protein 3, death associated protein 3, DKFZp686G12159,28S ribosomal protein S29, mitochondrial, FLJ12817, MGC126058, MGC126059, mitochondrial 28S ribosomal protein S29, MRP-S29Ionizing radiation resistance conferring protein, MRPS29S29mt | |
| DAP3 | |
| IgG | |
| Protein G purified | |
| In Jurkat, a 45 kDa band is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title