missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DAP10/HCST Antibody (982107), Unconjugated, Novus Biologicals™
Mouse Monoclonal Antibody
£178.00 - £375.00
Specifications
| Antigen | DAP10/HCST |
|---|---|
| Clone | 982107 |
| Concentration | 1.0 mg/mL |
| Dilution | Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 5 μg/mL, Immunohistochemistry-Paraffin 5 μg/mL |
| Applications | Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18612894
|
Novus Biologicals
MAB97862-SP |
25 μg |
£178.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18632345
|
Novus Biologicals
MAB97862 |
100 μg |
£375.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DAP10/HCST Monoclonal antibody specifically detects DAP10/HCST in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| DAP10/HCST | |
| 1.0 mg/mL | |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Adaptive Immunity, Immune System Diseases, Immunology, Innate Immunity | |
| PBS | |
| 10870 | |
| IgG2a | |
| Protein A or G purified |
| 982107 | |
| Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 5 μg/mL, Immunohistochemistry-Paraffin 5 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| DAP10PIK3AP, DKFZP586C1522, hematopoietic cell signal transducer, KAP10DNAX-activation protein 10, kinase assoc pro of ∽10kDa, kinase assoc protein, Membrane protein DAP10, phosphoinositide-3-kinase adaptor protein, Transmembrane adapter protein KAP10 | |
| Synthetic peptide containing human DAP10/HCST | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title