missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
DAP1 Polyclonal antibody specifically detects DAP1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | DAP1 |
| Applications | ELISA |
| Immunogen | DAP1 antibody was raised against a 19 amino acid peptide near the carboxy terminus of human DAP1. The immunogen is located within the last 50 amino acids of DAP1. Amino Acid Squence: ASMDKHPSPRTQHIQQPRK |
| Purification Method | Peptide affinity purified |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?