missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DAP1 Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-82082AF532
This item is not returnable.
View return policy
Description
DAP1 Polyclonal antibody specifically detects DAP1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| DAP1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 532 | |
| DAP1, DAP-1, death-associated protein, death-associated protein 1, MGC99796 | |
| DAP1 antibody was raised against a 19 amino acid peptide near the carboxy terminus of human DAP1. The immunogen is located within the last 50 amino acids of DAP1. Amino Acid Squence: ASMDKHPSPRTQHIQQPRK | |
| 0.1 mL | |
| Apoptosis, Cellular Markers | |
| 1611 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction