missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DAP Kinase 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38180-20ul
This item is not returnable.
View return policy
Description
DAP Kinase 1 Polyclonal antibody specifically detects DAP Kinase 1 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| DAP Kinase 1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:100 | |
| DAPKDAP kinase 1, death-associated protein kinase 1, DKFZp781I035, EC 2.7.11, EC 2.7.11.1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1141-1430 of human DAP Kinase 1 (DAPK1) (NP_004929.2).,, Sequence:, LLTVKHYLSPQQLREHHEPVMIYQPRDFFRAQTLKETSLTNTMGGYKESFSSIMCFGCHDVYSQASLGMDIHASDLNLLTRRKLSRLLDPPDPLGKDWCLLAMNLGLPDLVAKYNTSNGAPKDFLPSPLHALLREWTTYPESTVGTLMSKLRELGRRDAADFLLKASSVFKINLDGNGQEAYASSCNSGTSYNSISSVVSR | |
| 20 μL | |
| Apoptosis, Cancer, Phospho Specific | |
| 1612 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction