missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DAO Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA576708
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
This gene encodes the peroxisomal enzyme D-amino acid oxidase. The enzyme is a flavoprotein which uses flavin adenine dinucleotide (FAD) as its prosthetic group. Its substrates include a wide variety of D-amino acids, but it is inactive on the naturally occurring L-amino acids. Its biological function is not known; it may act as a detoxifying agent which removes D-amino acids that accumulate during aging. In mice, it degrades D-serine, a co-agonist of the NMDA receptor. This gene may play a role in the pathophysiology of schizophrenia.
Specifications
| DAO | |
| Polyclonal | |
| Unconjugated | |
| DAO | |
| ABP; Abp1; AI987963; AOC1; DAAO; D-amino acid oxidase; D-amino acid oxidase (EC 1.4.3.3); D-amino acid oxidase 1; D-amino-acid oxidase; DAMOX; Dao; DAO1; Dao-1; Diamine oxidase; Histaminase; KAO; OXDA | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 114027, 13142, 1610 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| O35078, P14920, P18894 | |
| DAO | |
| Recombinant full length Human DAO. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction