missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DAO Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33326-100ul
This item is not returnable.
View return policy
Description
DAO Monoclonal antibody specifically detects DAO in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| DAO | |
| Monoclonal | |
| Western Blot 1:2000 - 1:20000, ELISA Recommended starting concentration is 1 μg/mL | |
| DAAO, D-amino-acid oxidase, DAMOXOXDA, DAO, EC 1.4.3, EC 1.4.3.3, MGC35381 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human DAO (NP_001908.3).,, Sequence:, MRVVVIGAGVIGLSTALCIHERYHSVLQPLDIKVYADRFTPLTTTDVAAGLWQPYLSDPNNPQEADWSQQTFDYLLSHVHSPNAENLGLFLISGYNLFHE | |
| 100 μL | |
| Amino Acids Drugs and other small molecules, Endocrinology, Signal Transduction | |
| 1610 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction