missing translation for 'onlineSavingsMsg'
Learn More

Dantrolene sodium salt

Product Code. 15463579
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15463579 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15463579 Supplier Thermo Scientific Alfa Aesar Supplier No. J60887.MC
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Inhibits intracellular calcium release from the sarcoplasmic reticulum | CAS: 14663-23-1 | C14H12N4NaO5 | 339.263 g/mol

Inhibits intracellular calcium release from the sarcoplasmic reticulum

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14663-23-1
Molecular Formula C14H12N4NaO5
Molecular Weight (g/mol) 339.263
MDL Number MFCD00079130
InChI Key XHONPQLVCHTEDY-YVCISUPJSA-N
PubChem CID 131674048
IUPAC Name molecular hydrogen;1-[(E)-[5-(4-nitrophenyl)furan-2-yl]methylideneamino]imidazolidine-2,4-dione;sodium
SMILES [HH].C1C(=O)NC(=O)N1N=CC2=CC=C(O2)C3=CC=C(C=C3)[N+](=O)[O-].[Na]

Specifications

Color Orange to Yellow
Quantity 100 mg
Merck Index 14,2816
Formula Weight 336.23
Physical Form Crystalline Powder
Chemical Name or Material Dantrolene sodium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.