missing translation for 'onlineSavingsMsg'
Learn More

Danthron, MedChemExpress

Product Code. 16438516
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16438516 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16438516 Supplier Medchem Express Supplier No. HYB0923
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Danthron is a natural product extracted from the traditional Chinese medicine rhubarb. Danthron functions in regulating glucose and lipid metabolism by activating AMPK.

Danthron is a natural product extracted from the traditional Chinese medicine rhubarb. Danthron functions in regulating glucose and lipid metabolism by activating AMPK.

Specifications

Chemical Name or Material Danthron
CAS 117-10-2
Color Deep Yellow
Purity Grade Notes Research
Molecular Formula C14H8O4
Quantity 100 mg
Synonym Dantron Chrysazin 1,8-Dihydroxyanthraquinone
Solubility Information DMSO : 5 mg/mL (20.82 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C1C2=C(C=CC=C2O)C(C3=CC=CC(O)=C13)=O
Molecular Weight (g/mol) 240.21
Formula Weight 240.21
Percent Purity 97.72%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.