missing translation for 'onlineSavingsMsg'
Learn More

Dansylcadaverine, MedChemExpress

Product Code. 16444697
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16444697 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16444697 Supplier Medchem Express Supplier No. HYD102750mg
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Dansylcadaverine (Monodansyl cadaverine) is an autofluorescent compound used for the labeling of autophagic vacuoles. Dansylcadaverine, a high affinity substrate of transglutaminases, can block the receptor-mediated endocytosis of many ligands.

Dansylcadaverine (Monodansyl cadaverine) is an autofluorescent compound used for the labeling of autophagic vacuoles. Dansylcadaverine, a high affinity substrate of transglutaminases, can block the receptor-mediated endocytosis of many ligands.

Specifications

Chemical Name or Material Dansylcadaverine
CAS 10121-91-2
Purity Grade Notes Research
Molecular Formula C17H25N3O2S
Synonym Monodansyl cadaverine
Color White
Solubility Information DMSO : 62.5 mg/mL (186.31 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 80°C) (insoluble)
Quantity 50 mg
Physical Form Solid
SMILES NCCCCCNS(=O)(C1=CC=CC2=C1C=CC=C2N(C)C)=O
Molecular Weight (g/mol) 335.46
Formula Weight 335.46
Percent Purity 98.07%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.