missing translation for 'onlineSavingsMsg'
Learn More

Dansyl Chloride, TRC

Product Code. 17951955
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17951955 5 g 5g
17904545 250 mg 250mg
17913005 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17951955 Supplier Toronto Research Chemicals Supplier No. TRCD1775005G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 605-65-2
Molecular Formula C12 H12 Cl N O2 S
Molecular Weight (g/mol) 269.75
Synonym 1-Naphthalenesulfonyl chloride, 5-(dimethylamino)- (6CI, 7CI, 8CI, 9CI, ACI), 5-(Dimethylamino)-1-naphthalenesulfonyl chloride (ACI), 1-(Dimethylamino)-5-naphthalenesulfonyl chloride, 5-(N,N-Dimethylamino)-1-naphthalenesulfonyl chloride, 8-Dimethylamino-1-naphthalenesulfonyl chloride, Dansyl chloride, DNS chloride, DNS-Cl, NSC 83616
IUPAC Name 5-(dimethylamino)naphthalene-1-sulfonyl chloride
SMILES CN(C)c1cccc2c(cccc12)S(=O)(=O)Cl

Specifications

Quantity 5 g
InChI Formula InChI=1S/C12H12ClNO2S/c1-14(2)11-7-3-6-10-9(11)5-4-8-12(10)17(13,15)16/h3-8H,1-2H3
Formula Weight 269.0277
Percent Purity >95%
Chemical Name or Material Dansyl Chloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.