missing translation for 'onlineSavingsMsg'
Learn More

Dansyl Chloride (Crystalline (Yellow)), Fisher BioReagents

Product Code. 10541255
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10541255 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10541255 Supplier Fisher BioReagents Supplier No. BP4271

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Dansyl Chloride is a fluorochrome useful in the detection of N-terminal amino acids in proteins and peptides. It is also used in the preparation of fluorescent derivatives of amino acids.
TRUSTED_SUSTAINABILITY

Specifications

Melting Point 71°C
CAS 605-65-2
Assay Percent Range ≥99 %
Molecular Formula C12H12ClNO2S
MDL Number MFCD00003985
UN Number 1759
Merck Index 15, 2813
Synonym 1-(Dimethylamino)-5-naphthalenesulfonyl Chloride
Identification Pass Test
InChI Key XPDXVDYUQZHFPV-UHFFFAOYSA-N
SMILES CN(C)C1=CC=CC2=C(C=CC=C12)S(Cl)(=O)=O
IUPAC Name 5-(dimethylamino)naphthalene-1-sulfonyl chloride
Molecular Weight (g/mol) 269.74
PubChem CID 11801
ChEBI CHEBI:51907
Formula Weight 269.75
Percent Purity ≥99%
Packaging Amber Glass
Color Yellow
Quantity 1 g
Physical Form Powder
Chemical Name or Material Dansyl Chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.