missing translation for 'onlineSavingsMsg'
Learn More

Dansyl chloride, 98%

Product Code. 10348743 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10348743 25 g Glass bottle 25g
10540571 1 g Glass bottle 1g
10070880 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 10348743 Leverantör Thermo Scientific Acros Leverantörsnummer 115850250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

CAS: 605-65-2 | C12H12ClNO2S | 269.74 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 605-65-2
Molecular Formula C12H12ClNO2S
Molecular Weight (g/mol) 269.74
MDL Number MFCD00003985
InChI Key XPDXVDYUQZHFPV-UHFFFAOYSA-N
Synonym 5-Dimethylamino-1-naphthalenesulfonyl chloride
PubChem CID 11801
ChEBI CHEBI:51907
IUPAC Name 5-(dimethylamino)naphthalene-1-sulfonyl chloride
SMILES CN(C)C1=CC=CC2=C(C=CC=C12)S(Cl)(=O)=O

Specifikationer

Melting Point 71.0°C to 74.0°C
Assay Percent Range 98%
Solubility Information Solubility in water: 10g/L (20°C). Other solubilities: soluble in acetone, pyridine, benzene and dioxane
Infrared Spectrum Authentic
Formula Weight 269.75
Percent Purity 98%
Packaging Glass bottle
Color Beige/Brown or Gray to White
Quantity 25 g
Physical Form Powder, Needles or Crystals
Chemical Name or Material Dansyl chloride, 98%
Visa mer Visa mindre

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.