missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Danoprevir, MedChemExpress
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
Unit Size:
10mg
2mg
50mg
5mg
Description
Danoprevir (ITMN-191) is an orally active NS3/4A protease inhibitor for hepatitis C virus (HCV) with an IC50 of 0.29 nM and is selective for NS3/4A over a panel of 53 proteases (IC50 higher than 10 μM). Danoprevir (ITMN-191) inhibits HCV genotypes 1a, 1b, 4, 5, and 6 (IC50s=0.2-0.4 nM) as well as 2b and 3a (IC50s=1.6, 3.5 nM). Danoprevir is also a SARS-CoV 3CLpro inhibitor with an IC50 of 0.05 μM.
Specifications
Specifications
| Chemical Name or Material | Danoprevir |
| CAS | 850876-88-9 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C35H46FN5O9S |
| Quantity | 50 mg |
| Synonym | ITMN-191 R7227 RO5190591 RG7227 |
| Solubility Information | DMSO : ≥ 100 mg/mL (136.64 mM) |
| SMILES | O=S(NC([C@@]1(NC([C@]2([H])C[C@@H](OC(N3CC4=C(C3)C=CC=C4F)=O)CN2C5=O)=O)C[C@]1(/C=C/CCCCC[C@@H]5NC(OC(C)(C)C)=O)[H])=O)(C6CC6)=O |
| Molecular Weight (g/mol) | 731.83 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction