missing translation for 'onlineSavingsMsg'
Learn More

Danofloxacin Mesylate, TRC

Product Code. 30009934
Change view
Click to view available options
Quantity:
10 mg
100 mg
1 g
Unit Size:
100mg
10mg
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009934 100 mg 100mg
30005262 1 g 1g
30003994 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009934 Supplier Toronto Research Chemicals Supplier No. TRCD153400100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119478-55-6
Molecular Formula C19 H20 F N3 O3 . C H4 O3 S
Molecular Weight (g/mol) 453.48
Synonym 3-Quinolinecarboxylic acid, 1-cyclopropyl-6-fluoro-1,4-dihydro-7-(5-methyl-2,5-diazabicyclo[2.2.1]hept-2-yl)-4-oxo-, (1S)-, monomethanesulfonate, 3-Quinolinecarboxylic acid, 1-cyclopropyl-6-fluoro-1,4-dihydro-7-[(1S,4S)-5-methyl-2,5-diazabicyclo[2.2.1]hept-2-yl]-4-oxo-, monomethanesulfonate (9CI), 2,5-Diazabicyclo[2.2.1]heptane, 3-quinolinecarboxylic acid deriv., CP 76136-27, Danofloxacin mesylate
IUPAC Name 1-cyclopropyl-6-fluoro-7-[(1S,4S)-5-methyl-2,5-diazabicyclo[2.2.1]heptan-2-yl]-4-oxoquinoline-3-carboxylic acid;methanesulfonic acid
SMILES CN1C[C@@H]2C[C@H]1CN2c3cc4N(C=C(C(=O)O)C(=O)c4cc3F)C5CC5.CS(=O)(=O)O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C19H20FN3O3.CH4O3S/c1-21-7-12-4-11(21)8-22(12)17-6-16-13(5-15(17)20)18(24)14(19(25)26)9-23(16)10-2-3-10;1-5(2,3)4/h5-6,9-12H,2-4,7-8H2,1H3,(H,25,26);1H3,(H,2,3,4)/t11-,12-;/m0./s1
Formula Weight 453.137
Chemical Name or Material Danofloxacin mesylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.