missing translation for 'onlineSavingsMsg'
Learn More

Danofloxacin mesylate, MedChemExpress

Product Code. 16470958
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16470958 5 mg 5mg
16468508 10 mg 10mg
16464377 50 mg 50mg
16448346 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16470958 Supplier Medchem Express Supplier No. HYB0501

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Danofloxacin mesylate (CP 76136-27) is a fluoroquinolone antibacterial for veterinary use.

Danofloxacin mesylate (CP 76136-27) is a fluoroquinolone antibacterial for veterinary use.

Specifications

Chemical Name or Material Danofloxacin mesylate
Physical Form Solid
CAS 119478-55-6
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C20H24FN3O6S
For Use With (Application) COVID-19-immunoregulation
Synonym CP 76136-27
Solubility Information H2O : 100 mg/mL (220.52 mM; Need ultrasonic) ∣DMSO : 20 mg/mL (44.10 mM; Need ultrasonic)
SMILES O=C(C1=CN(C2CC2)C3=C(C=C(F)C(N4[C@](C5)([H])CN(C)[C@]5([H])C4)=C3)C1=O)O.CS(=O)(O)=O
Molecular Weight (g/mol) 453.48
Formula Weight 453.48
Percent Purity 99.81%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.