missing translation for 'onlineSavingsMsg'
Learn More

Danofloxacin Mesilate, Mikromol™

Product Code. 17932466
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17932466 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17932466 Supplier Mikromol Supplier No. MM3710.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Danofloxacin Mesilate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Danofloxacin mesylate
CAS 119478-55-6
Analyte or Component Names Danofloxacin mesylate
Shipping Condition Room Temperature
Molecular Formula C19 H20 F N3 O3 . C H4 O3 S
Certifications/Compliance ISO 17034
Synonym 3-Quinolinecarboxylic acid, 1-cyclopropyl-6-fluoro-1,4-dihydro-7-[(1S,4S)-5-methyl-2,5-diazabicyclo[2.2.1]hept-2-yl]-4-oxo-, methanesulfonate (1:1), 3-Quinolinecarboxylic acid, 1-cyclopropyl-6-fluoro-1,4-dihydro-7-(5-methyl-2,5-diazabicyclo[2.2.1]hept-2-yl)-4-oxo-, (1S)-, monomethanesulfonate, 3-Quinolinecarboxylic acid, 1-cyclopropyl-6-fluoro-1,4-dihydro-7-[(1S,4S)-5-methyl-2,5-diazabicyclo[2.2.1]hept-2-yl]-4-oxo-, monomethanesulfonate (9CI), 2,5-Diazabicyclo[2.2.1]heptane, 3-quinolinecarboxylic acid deriv., CP 76136-27, Danofloxacin mesylate
InChI Formula InChI=1S/C19H20FN3O3.CH4O3S/c1-21-7-12-4-11(21)8-22(12)17-6-16-13(5-15(17)20)18(24)14(19(25)26)9-23(16)10-2-3-10;1-5(2,3)4/h5-6,9-12H,2-4,7-8H2,1H3,(H,25,26);1H3,(H,2,3,4)/t11-,12-;/m0./s1
SMILES CN1C[C@@H]2C[C@H]1CN2c3cc4N(C=C(C(=O)O)C(=O)c4cc3F)C5CC5.CS(=O)(=O)O
IUPAC Name 1-cyclopropyl-6-fluoro-7-[(1S,4S)-5-methyl-2,5-diazabicyclo[2.2.1]heptan-2-yl]-4-oxoquinoline-3-carboxylic acid;methanesulfonic acid
Molecular Weight (g/mol) 453.48
Quantity 250 mg
Formula Weight 453.137 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.