missing translation for 'onlineSavingsMsg'
Learn More

Manutan Danger hazardous area A board sign, 590mm x 290mm

Product Code. 12871749
Encompass_Preferred
This item is not returnable. View return policy

Product Code. 12871749

Brand: Manutan A346044

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
A raneg of double sided fluted polypropylene display boards that are lightweight and easy to assemble. Can be folded flat for storage and is easy to clean.
  • Danger hazardous area
  • Dimensions (H x W) : 590mm x 290mm
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.