missing translation for 'onlineSavingsMsg'
Learn More

Manutan Danger biohazard sign, self adhesive vinyl, 150mm x 200mm

Product Code. 12896378
Encompass_Preferred
This item is not returnable. View return policy

Product Code. 12896378

Brand: Manutan A347119

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
Ideal for low-light areas, warn of immediately hazardous situations.
  • Photoluminescent self-adhesive vinyl
  • Dimensions (H x W) : 150mm x 200mm
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.