missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DAND5 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22137-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the BMP (bone morphogenic protein) antagonist family. Like BMPs, BMP antagonists contain cystine knots and typically form homo- and heterodimers. The CAN (cerberus and dan) subfamily of BMP antagonists, to which this gene belongs, is characterized by a C-terminal cystine knot with an eight-membered ring. The antagonistic effect of the secreted protein encoded by this gene is likely due to its direct binding to BMP proteins. As an antagonist of BMP, this gene may play a role in regulating organogenesis, body patterning, and tissue differentiation. In mouse, this protein has been shown to bind Nodal and to inhibit the Nodal signaling pathway which patterns left/right body asymmetry.Specifications
| DAND5 | |
| Polyclonal | |
| Unconjugated | |
| DAND5 | |
| CER2, Cerberus like protein 2, Cerl 2, CERL2, CKTSF1B3, COCO, CRL2, DAN domain family member 5, DAN domain family, member 5, DAND5, DANTE, GREM3, Gremlin 3, SP1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 199699 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.36 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8N907 | |
| DAND5 | |
| DAND5 Fusion Protein Ag17533 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction